Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant IL-7 (from Yeast)
Human Recombinant IL-7 (from Yeast)
Supplier:  Apollo Scientific
Certificates
Human Recombinant IL-7 (from Yeast)
Supplier:  Apollo Scientific

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Yeast
  • Species:
    Human
  • Protein synonyms:
    Interleukin-7
  • Protein/peptide name:
    IL-7

Specifications

Frequently Bought Together