0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteins and Peptides
Human recombinant IFN-a1
Human recombinant IFN-a1
Human recombinant IFN-a1
Catalog # APOSBITP1092-5UG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Human recombinant IFN-a1
Catalog # APOSBITP1092-5UG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
5 g
Protein/peptide type:
Recombinant
Species:
Human
Seq:
03
Protein synonyms:
Interferon-alpha 1
Protein/peptide name:
IFN-a1
Specifications
About this item
Product Details
Product Details