Mouse recombinant GM-CSF
Catalog # APOSBITP1086-5UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:5 g
- Protein/peptide type:Recombinant
- Source:
- Species:Mouse
- Seq:03
- Environmentally Preferable:
- Protein synonyms:Granulocyte Macrophage-Colony Stimulating Factor
- Protein/peptide name:GM-CSF