Order Entry
Czech Republic
ContactUsLinkComponent
 
Human recombinant MIP-3b
Catalog # APOSBITC2038-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant MIP-3b
Catalog # APOSBITC2038-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Macrophage Inflammatory protein-3 beta, CCL19
  • Protein/peptide name:
    MIP-3b

Specifications

About this item