Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Proteins and Peptides
Human recombinant IP-10
Human recombinant IP-10
Human recombinant IP-10
Catalog # APOSBITC2018-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Human recombinant IP-10
Catalog # APOSBITC2018-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
CXCL10
Protein/peptide name:
IP-10
Specifications
About this item
Product Details
Product Details