Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Mouse recombinant C-10
Mouse recombinant C-10
Mouse recombinant C-10
Catalog # APOSBITC2000-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Mouse recombinant C-10
Catalog # APOSBITC2000-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Mouse
Protein synonyms:
CCL6
Protein/peptide name:
C-10
Specifications
About this item
Product Details
Product Details