Order Entry
Czech Republic
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Tests, Oxygen
NANOCOLOR® Photometric Tube Tests, Oxygen
Catalog # MANA985082
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Tests, Oxygen
Catalog # MANA985082
Supplier:  MACHEREY-NAGEL
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Chemical method:
    Winkler
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Oxygen 12, test no. 0-82
  • Dimensions:
    160×110×250 mm (Including packaging)
  • Model:
    MANA985082
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    24
  • Weight:
    490 g (Including packaging)
  • For unit:
    UV/VIS II;VIS II;500 D;PF-12Plus;Advance;MT Easy VIS;MT Easy UV
  • Tests per kit:
    22
  • Range:
    0,5 - 12,0 mg/L ((O₂))

Specifications

About this item

Photometric test kit for the determination of oxygen in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Colored pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (AD) tubes. It is pre-programmed on NANOCOLOR® photometers and is also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: Yes

Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

Sludge reagent: No

Digestion: No

NanOx metal: No

NanOx: No

Delivery:

Rugged box with 22 test tubes. Sufficient for 22 tests.