NANOCOLOR® Photometric Tube Test Kit for Organic Complexing Agents
MANA985052
:
- Chemical method:Bismut xylenol orange
- Pk:1 KIT
- Description:NANOCOLOR® organic Complexing Agents 10, test no. 0 - 52
- Dimensions:160×110×119 mm (Including packaging)
- Model:MANA985052
- Platform:NANOCOLOR® tube tests
- Storage temperature:15...25 °C
- Shelf life (months):12
- Weight:338 g (Including packaging)
- For unit:UV/VIS II;VIS II;500 D;PF-12Plus;Advance
- Tests per kit:19
- Range:0.5 - 15.0 mg/L - IBiK
Photometric test kit for the determination of organic complexing agents in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Colored pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (AD) round cells. It is pre-programmed on the NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: No
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
: Rugged box with 19 test tubes. Sufficient for 10-19 tests.