Tat-NR2B9c ≥98%
Supplier: Apollo Scientific
Jump to:
About this item
Tat-NR2B9c is a PSD95 inhibitor with EC50 of 7 nM, also inhibit p38.
- CAS Number:500992-11-0
- Formula:C₁₀₅H₁₈₈N₄₂O₃₀
- MDL Number:MFCD09842404
- Molecular Weight:2518.88 g/mol
- Storage Temperature:Freezer
3 Options Available Below
Product Details & Documents
Tat-NR2B9c is a PSD95 inhibitor with EC50 of 7 nM, also inhibit p38.
- CAS Number:500992-11-0
- Formula:C₁₀₅H₁₈₈N₄₂O₃₀
- MDL Number:MFCD09842404
- Molecular Weight:2518.88 g/mol
- Storage Temperature:Freezer
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



