VWR®, Precision Grip Gloves, NBR Coating
Supplier: VWR Collection
Certificates
About this item
This VWR® precision grip glove (with nitrile coating) provides basic protection against mechanical risks.
- Nylon composition
- Nitrile (NBR) coating on palm and fingertips
- Blue knitted glove with black coating
Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment. Also great for package handling, picking and packing.
Certifications: Meets requirements for EN 388:2016 (4131X). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications
- Catalog No:
- 112-5738
- 112-5735
- 112-5737
- 112-5739
- 112-5736
- 112-5740
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Finish:
- Nitrile palm coating
- Nitrile palm coating
- Nitrile palm coating
- Nitrile palm coating
- Nitrile palm coating
- Nitrile palm coating
- Gauge:
- 15
- 15
- 15
- 15
- 15
- 15
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
Specifications
Related Information
VWR® Precision Grip Gloves, NBR Coating
Additional Product Details
VWR® Reusable Cut Resistant and Precision Work Glove Guide
Product Literature
Frequently Bought Together