Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Nivalenol standard solution

Nivalenol standard solution

Supplier:   Apollo Scientific
Nivalenol standard solution
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents

Analytical reference standard solution in acetonitrile.

Caution: Expires in 12 months.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • APOSBIMS126-1ML
  • APOSBIMS126-5ML
  • Description:
  • Nivalenol
  • Nivalenol
  • CAS No.:
  • 23282-20-4
  • 23282-20-4
  • Formula:
  • C₁₅H₂₀O₇
  • C₁₅H₂₀O₇
  • Flash point:
  • 2 °C
  • 2 °C
  • Storage temperature:
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Concentration
    Pack type
    Pk
    Availability
    Price
  • Nivalenol
    100 µg/ml in acetonitrile
    Bottle
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 7 180,00 Kč
    7 180,00 Kč
    1 * 1 mL
       7 180,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIMS126-1ML
    Description
    Nivalenol
    CAS No.
    23282-20-4
    Formula
    C₁₅H₂₀O₇
    Flash point
    2 °C
    Storage temperature
    Freezer
  • Nivalenol
    100 µg/ml in acetonitrile
    Bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 28 080,00 Kč
    28 080,00 Kč
    1 * 5 mL
       28 080,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIMS126-5ML
    Description
    Nivalenol
    CAS No.
    23282-20-4
    Formula
    C₁₅H₂₀O₇
    Flash point
    2 °C
    Storage temperature
    Freezer
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.