Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Zearalenone standard solution

Zearalenone standard solution

Supplier:   Apollo Scientific
Zearalenone standard solution
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents

Analytical reference standard solution in acetonitrile.

Caution: Expires in 12 months.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • APOSBIMS119-1ML
  • APOSBIMS119-5ML
  • Description:
  • Zearalenone
  • Zearalenone
  • CAS No.:
  • 17924-92-4
  • 17924-92-4
  • Formula:
  • C₁₈H₂₂O₅
  • C₁₈H₂₂O₅
  • Boiling point:
  • 81…82 °C
  • 81…82 °C
  • Flash point:
  • 2 °C
  • 2 °C
  • Storage temperature:
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Concentration
    Pack type
    Pk
    Availability
    Price
  • Zearalenone
    100 µg/ml in acetonitrile
    Bottle
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 6 150,00 Kč
    6 150,00 Kč
    1 * 1 mL
       6 150,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIMS119-1ML
    Description
    Zearalenone
    CAS No.
    17924-92-4
    Formula
    C₁₈H₂₂O₅
    Boiling point
    81…82 °C
    Flash point
    2 °C
    Storage temperature
    Freezer
  • Zearalenone
    100 µg/ml in acetonitrile
    Bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 24 390,00 Kč
    24 390,00 Kč
    1 * 5 mL
       24 390,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIMS119-5ML
    Description
    Zearalenone
    CAS No.
    17924-92-4
    Formula
    C₁₈H₂₂O₅
    Boiling point
    81…82 °C
    Flash point
    2 °C
    Storage temperature
    Freezer
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.