Order Entry
Czech Republic
ContactUsLinkComponent
 
Ochratoxin A standard solution
 
Ochratoxin A standard solution
 

 

  • CAS No.:
    303-47-9
  • Formula:
    C₂₀H₁₈ClNO₆
  • Storage temperature:
    Freezer
  • Description:
    Ochratoxin A
  • Flash point:
    2 °C

 

 

Frequently Bought Together