Fumonisin Standard Solutions
Analytical reference standard solutions derived from Fusarium moniliforme. Also known as Macrofusine
- 50µg/ml of each component in ACN/H2O 1:1 V/V mixture.
Carcinogenic mycotoxin.
: Expires in 12 months.
- CAS No.:116355-84-1
- Formula:C₃₄H₅₉NO₁₄
- Storage temperature:Freezer
- Description:Fumonisin B2
Frequently Bought Together