NANOCOLOR® Photometric Tube Tests, Cadmium
Katalogové číslo MANA985014
Dodavatel: MACHEREY-NAGEL
Číslo CAS:
Specifikace
- Chemical method:Cadion
- Pk:1 KIT
- Description:NANOCOLOR® Cadmium 2, test no.0-14
- Dimensions:160×110×119 mm (including packaging)
- Seq:0
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:356 g (including packaging)
- Type:Hazardous material: Yes
- For unit:UV/VIS II; VIS II; 500 D; PF-12Plus; Advance; MT Easy VIS; MT Easy UV
- Tests per kit:19
- Range:0,05 - 2,00 mg/L (Cd²⁺)
Specifikace
O tomto produktu
Photometric test kit for the determination of cadmium in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Colored pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (AD) tubes. It is pre-programmed on NANOCOLOR® photometers and is also compatible with METTLER TOLEDO easy VIS and easy UV. Programming data for other spectrophotometers are available.
Sludge reagent set: No
Digestion set: Yes
NanOx Metal: Yes
NanOx N: No
Digestion temperature:
Delivery: Remark: Measuring range on NANOCOLOR VIS II