0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
2,6-Difluoro-4-iodoanisole
2,6-Difluoro-4-iodoanisole
2,6-Difluoro-4-iodoanisole
2,6-Difluoro-4-iodoanisole
APOSPC9917-1G
:
Apollo Scientific
:
2,6-Difluoro-4-iodoanisole
APOSPC9917-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
270.02 g/mol
:
GHS07
:
Ambient
:
886762-68-1
:
C₇H₅F₂IO
:
MFCD07368787