Order Entry
Czech Republic
ContactUsLinkComponent
5,6,7,8-Tetrafluorobenzo-1,4-dioxene 97+%
Irritant
5,6,7,8-Tetrafluorobenzo-1,4-dioxene 97+%
Catalog # APOSPC9876-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5,6,7,8-Tetrafluorobenzo-1,4-dioxene 97+%
Catalog # APOSPC9876-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt:78…79 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1743-87-9
Formula:C₈H₄F₄O₂
MDL Number:MFCD00229993