5'-Fluoro-2'-methoxyacetophenone 98%
Catalog # APOSPC9857-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:168.17 g/mol
Storage Temperature:Ambient
CAS Number:445-82-9
Formula:C₉H₉FO₂
MDL Number:MFCD00671763