
2,6-Difluoro-4-nitrophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:658-07-1
:
Formula:C₆H₃F₂NO₃
MDL Number:MFCD07366521
:175.0900 g/mol
Melting Pt:99.7…100.7 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together