

2-(Trifluoromethyl)pyrrolidine
Catalog # APOSPC9837-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:139.12 g/mol
Boiling Pt:110…112 °C
Density:1.21 g/cm³ (25 °C)
Flash Pt:26 °C
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Freezer
CAS Number:109074-67-1
Formula:C₅H₈F₃N
MDL Number:MFCD02663405