Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Difluoropropionic acid 97%
Irritant
2,2-Difluoropropionic acid 97%
Catalog # APOSPC9834-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,2-Difluoropropionic acid 97%
Catalog # APOSPC9834-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :110.06 g/mol
Boiling Pt:140…141 °C
Melting Pt:36…45 °C
Flash Pt:65 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:373-96-6
Formula:C₃H₄F₂O₂
MDL Number:MFCD03093764