Order Entry
Czech Republic
ContactUsLinkComponent
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid 97%
Irritant
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid 97%
Catalog # APOSPC9763-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid 97%
Catalog # APOSPC9763-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :275.23 g/mol
Melting Pt:140…142 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:465514-39-0
Formula:C₁₁H₁₂F₃N₃O₂
MDL Number:MFCD02091615