
trans-3-(Trifluoromethyl)cinnamoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64379-91-5
:
Formula:C₁₀H₆ClF₃O
MDL Number:MFCD00075514
:234.602 g/mol
Boiling Pt:148 °C
Density:1.372 g/cm³ (25 °C)
Flash Pt:94 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together