Order Entry
Czech Republic
ContactUsLinkComponent
1-[3-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Corrosive
Irritant
1-[3-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Catalog # APOSPC9582-100MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
1-[3-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Catalog # APOSPC9582-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :245.249 g/mol
Boiling Pt:90 °C/0.2mmHg
Storage Temperature:Ambient
UN:3267
ADR:8,III
 :GHS05
 :GHS07
CAS Number:243666-15-1
Formula:C₁₁H₁₄F₃N₃
MDL Number:MFCD12756231