
3-Methoxy-5-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56723-86-5
:
Formula:C₉H₇F₃O₂
MDL Number:MFCD08458144
:204.15 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together