
3-Fluoro-4-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:216144-68-2
:
Formula:C₈H₃ClF₄O
MDL Number:MFCD00236284
:226.56 g/mol
Boiling Pt:151…153 °C
Density:1.486 g/cm³ (25 °C)
Flash Pt:81 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together