4-Hydroxy-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:125647-79-2
:
Formula:C₁₁H₅F₆NO₂
MDL Number:MFCD00276584
:297.16 g/mol
Melting Pt:229…232 °C
Storage Temperature:Ambient
Frequently Bought Together