
4-Fluorobenzylhydrazine hydrochloride
Catalog # APOSPC9518-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:176.62 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
CAS Number:1059626-05-9
Formula:C₇H₁₀ClFN₂
MDL Number:MFCD08458859