6-{[3-(Trifluoromethyl)phenyl]sulphanyl}imidazo[2,1-b][1,3]thiazole-5-carbaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175277-56-2
:
Formula:C₁₃H₇F₃N₂OS₂
MDL Number:MFCD00102587
Melting Pt:86…89 °C
:328.34 g/mol
Storage Temperature:Ambient
Frequently Bought Together