Methyl-4-chloro-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-pyrrolidinecarboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:251310-42-6
:
Formula:C₁₂H₁₁Cl₂F₃N₂O₂
MDL Number:MFCD00664157
:343.13 g/mol
Melting Pt:84…87 °C
Storage Temperature:Ambient
Frequently Bought Together