Order Entry
Czech Republic
ContactUsLinkComponent
6-(4-Fluorophenyl)-2-(methylsulphanyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Irritant
6-(4-Fluorophenyl)-2-(methylsulphanyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Catalog # APOSPC9359-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-(4-Fluorophenyl)-2-(methylsulphanyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Catalog # APOSPC9359-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :275.31 g/mol
Melting Pt:286…288 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:338394-83-5
Formula:C₁₂H₁₀FN₅S
MDL Number:MFCD00138905