
2-Hydroxy-6-(trifluoromethyl)pyridine
Catalog # APOSPC9263-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:163.1 g/mol
Boiling Pt:220 °C
Melting Pt:124…126 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
CAS Number:34486-06-1
Formula:C₆H₄F₃NO
MDL Number:MFCD08061317