
3-(Trifluoromethoxy)thiophenol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:220239-66-7
:
Formula:C₇H₅F₃OS
MDL Number:MFCD01320799
:194.18 g/mol
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together