
2,2-Difluorocyclopropylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:105614-25-3
:
Formula:C₃H₆ClF₂N
MDL Number:MFCD07777150
:129.54 g/mol
Melting Pt:145 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together