
3-(Difluoromethoxy)benzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4837-19-8
:
Formula:C₈H₆F₂O₃
MDL Number:MFCD00236224
:188.13 g/mol
Melting Pt:105…108 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together