
1,5-Dibromo-2,4-difluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:28342-75-8
:
Formula:C₆H₂Br₂F₂
MDL Number:MFCD08458122
Melting Pt:29…31 °C
:271.8849 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together