Order Entry
Czech Republic
ContactUsLinkComponent
5-Fluoro-2-mercaptobenzothiazole 98%
5-Fluoro-2-mercaptobenzothiazole 98%
Catalog # APOSPC9090-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Fluoro-2-mercaptobenzothiazole 98%
Catalog # APOSPC9090-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :185.25 g/mol
Storage Temperature:Ambient
CAS Number:155559-81-2
Formula:C₇H₄FNS₂
MDL Number:MFCD02181177