cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1202578-27-5
:
Formula:C₈H₁₁F₃O₂
MDL Number:MFCD09037361
:196.17 g/mol
Storage Temperature:Ambient
Frequently Bought Together