2,5-difluoro-3-nitrotoluene 97%
Katalogové číslo APOSPC906352-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:173.119 g/mol
Storage Temperature:Ambient
CAS Number:1093758-82-7
Formula:C₇H₅F₂NO₂
MDL Number:MFCD11519028