

3-(Perfluorohexyl)propanol-1 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:80806-68-4
:
Formula:C₉H₇F₁₃O
MDL Number:MFCD00077578
:378.13 g/mol
Boiling Pt:70…80 °C/10mmHg
Melting Pt:–3 °C
Density:1.629 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1987
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together