Order Entry
Czech Republic
ContactUsLinkComponent
[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol 97%
[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol 97%
Catalog # APOSPC902881-1G
Supplier:  Apollo Scientific
CAS Number:  
[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol 97%
Catalog # APOSPC902881-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.19 g/mol
Storage Temperature:Ambient
CAS Number:153863-34-4
Formula:C₁₀H₉FN₂O
MDL Number:MFCD08059807