Order Entry
Czech Republic
ContactUsLinkComponent
5-[3,5-Bis(trifluoromethyl)phenyl]-1,2,4-triazole-3-(2H)-thione 97%
Irritant
5-[3,5-Bis(trifluoromethyl)phenyl]-1,2,4-triazole-3-(2H)-thione 97%
Catalog # APOSPC9025-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-[3,5-Bis(trifluoromethyl)phenyl]-1,2,4-triazole-3-(2H)-thione 97%
Catalog # APOSPC9025-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:280 °C
 :313.23 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:175276-77-4
Formula:C₁₀H₅F₆N₃S
MDL Number:MFCD00052515