
5,7-Bis(trifluoromethyl)-2-(methylthio)pyrazolo[1,5-a]pyrimidine-3-carboxamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175203-36-8
:
Formula:C₁₀H₆F₆N₄OS
MDL Number:MFCD00174394
:344.24 g/mol
Melting Pt:>250 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together