0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
3-Iodo-2-(trifluoromethyl)pyridine
3-Iodo-2-(trifluoromethyl)pyridine
3-Iodo-2-(trifluoromethyl)pyridine
3-Iodo-2-(trifluoromethyl)pyridine
APOSPC8980-1G
:
Apollo Scientific
:
3-Iodo-2-(trifluoromethyl)pyridine
APOSPC8980-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
273 g/mol
:
42…43 °C
:
GHS07
:
Ambient
:
590371-71-4
:
C₆H₃F₃IN
:
MFCD08234938