
4-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:914636-59-2
:
Formula:C₁₂H₇F₃N₂O₃
MDL Number:MFCD08235203
:284.2 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together