
2,6-Dichloro-4-(trifluoromethyl)nicotinaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:914636-56-9
:
Formula:C₇H₂Cl₂F₃NO
MDL Number:MFCD08056286
:244 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together