Order Entry
Czech Republic
ContactUsLinkComponent
1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol
1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol
Catalog # APOSPC8926-500MG
Supplier:  Apollo Scientific
CAS Number:  
1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol
Catalog # APOSPC8926-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :287.21 g/mol
Melting Pt:65…67 °C
Storage Temperature:Ambient
CAS Number:478068-12-1
Formula:C₁₁H₁₁F₆NO
MDL Number:MFCD01871640