1,3-Di-boc-2-(trifluoromethylsulphonyl)guanidine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:207857-15-6
:
Formula:C₁₂H₂₀F₃N₃O₆S
MDL Number:MFCD02683528
:391.36 g/mol
Melting Pt:113 °C
Storage Temperature:Ambient
Frequently Bought Together