
4-Bromo-2-fluoro-5-iodotoluene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:870704-15-7
:
Formula:C₇H₅BrFI
MDL Number:MFCD06200719
:314.92
Boiling Pt:249…250 °C
Density:2.145 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together