
4-(1-Hydroxypropyl)benzotrifluoride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:67081-98-5
:
Formula:C₁₀H₁₁F₃O
MDL Number:MFCD07773046
:204.19 g/mol
Density:1.2 g/cm³ (25 °C)
Flash Pt:104 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together